Product Details

SNP ID
rs1610096
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1067823 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCTTAGGCAATCTGGGGGCTCT[A/G]GGGGCCGTTGAGATGGGAGGGCTGT
Phenotype
MIM: 605414 MIM: 601155
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ABCA7 PubMed Links

Gene Details

Gene
ABCA7
Gene Name
ATP binding cassette subfamily A member 7
There are no transcripts associated with this gene.

Gene
ARHGAP45
Gene Name
Rho GTPase activating protein 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258328.2 Intron NP_001245257.1
NM_001282334.1 Intron NP_001269263.1
NM_001282335.1 Intron NP_001269264.1
NM_001321232.1 Intron NP_001308161.1
NM_012292.4 Intron NP_036424.2
XM_006722713.1 Intron XP_006722776.1
XM_011527858.1 Intron XP_011526160.1
XM_011527860.2 Intron XP_011526162.1

View Full Product Details