Product Details

SNP ID
rs1621398
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:28660960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATCCATGTGTGATAGGCTGGGCT[A/G]GAGTGGTATAGTGAAGAAACCCTGC
Phenotype
MIM: 613386
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POMP PubMed Links

Gene Details

Gene
POMP
Gene Name
proteasome maturation protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015932.5 Intron NP_057016.1

View Full Product Details