Product Details

SNP ID
rs7396705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:9139974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTGAAGCTGGAGAAGCTGACAGC[C/T]CCCCACACCCCCCTTTCCCAAACAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DENND5A PubMed Links

Gene Details

Gene
DENND5A
Gene Name
DENN domain containing 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243254.1 Intron NP_001230183.1
NM_015213.3 Intron NP_056028.2
XM_005252832.1 Intron XP_005252889.1
XM_011519953.2 Intron XP_011518255.1
XM_017017419.1 Intron XP_016872908.1

View Full Product Details