Product Details

SNP ID
rs11037444
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5440855 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGCATGGCTCTCAGGATCAATGC[A/G]TAGGAAAGCAGGATGAAAGTTGAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR51B5 PubMed Links

Gene Details

Gene
OR51B5
Gene Name
olfactory receptor family 51 subfamily B member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005567.2 699 Intron NP_001005567.2
Gene
OR51I1
Gene Name
olfactory receptor family 51 subfamily I member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005288.2 699 Silent Mutation TAC,TAT Y220Y NP_001005288.1

View Full Product Details