Product Details

SNP ID
rs7129909
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5689947 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCATAGAATGGGAGGCTTTGAGAAG[A/G]ATAGATTGATTGTGTCTGGGTGAGT
Phenotype
MIM: 606559 MIM: 608487
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM22 PubMed Links

Gene Details

Gene
TRIM22
Gene Name
tripartite motif containing 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199573.1 Intron NP_001186502.1
NM_006074.4 Intron NP_006065.2
Gene
TRIM5
Gene Name
tripartite motif containing 5
There are no transcripts associated with this gene.

View Full Product Details