Product Details
- SNP ID
-
rs7990536
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:61409439 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAATACAAATCTAAAAGAACTTGG[C/T]TGTTTGCATTCAATGTCTTTTTTGT
- Phenotype
-
MIM: 614449
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PCDH20
PubMed Links
Gene Details
- Gene
- PCDH20
- Gene Name
- protocadherin 20
View Full Product Details