Product Details
- SNP ID
-
rs8100017
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:1451275 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGCTTCGGTCAGCTGAGAAGAACCA[C/T]GTGGGGGAGGCCTGGGTTTGGGGAG
- Phenotype
-
MIM: 612034
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
APC2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs8100440] are located under a probe and SNP(s) [rs8100004] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- APC2
- Gene Name
- APC2, WNT signaling pathway regulator
View Full Product Details