Product Details

SNP ID
rs265297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1452816 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTGCGCCCCGGATTGCCTGGCCA[C/G]CACCACGTGGGCCTGTACTTGTCCA
Phenotype
MIM: 612034
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
APC2 PubMed Links

Gene Details

Gene
APC2
Gene Name
APC2, WNT signaling pathway regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005883.2 Intron NP_005874.1
XM_005259475.2 Intron XP_005259532.1
XM_006722607.2 Intron XP_006722670.1
XM_006722608.3 Intron XP_006722671.2
XM_006722609.3 Intron XP_006722672.1
XM_006722610.3 Intron XP_006722673.2

View Full Product Details