Product Details

SNP ID
rs10071151
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:172645594 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGGCCCATGGAGTGATATTGCCT[C/T]TTCTCTTTCTACCTCTCCTCCCCCT
Phenotype
MIM: 615893
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NEURL1B PubMed Links

Gene Details

Gene
NEURL1B
Gene Name
neuralized E3 ubiquitin protein ligase 1B
There are no transcripts associated with this gene.

View Full Product Details