Product Details

SNP ID
rs10814993
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:4497428 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAATTCAGGAACCCTAAGGGAAGGA[A/C]GTTTGTGTTGCTCTTCTCTGTGCCC
Phenotype
MIM: 133550
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC1A1 PubMed Links

Gene Details

Gene
SLC1A1
Gene Name
solute carrier family 1 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004170.5 Intron NP_004161.4
XM_011518007.1 Intron XP_011516309.1
XM_011518008.2 Intron XP_011516310.1
XM_011518009.2 Intron XP_011516311.1
XM_011518010.1 Intron XP_011516312.1
XM_017015042.1 Intron XP_016870531.1
XM_017015043.1 Intron XP_016870532.1

View Full Product Details