Product Details
- SNP ID
-
rs7871596
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:69042481 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTGAGTACCACGATCCCTTCCTTCA[A/G]AGGAGTAAACTGAGGTTCCAAAAGG
- Phenotype
-
MIM: 606829
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FXN
PubMed Links
Gene Details
- Gene
- FXN
- Gene Name
- frataxin
View Full Product Details