Product Details

SNP ID
rs10807485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:54854893 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAATATTCAACAATATGTTTCTA[A/G]TATTTTATACTTTTAAAAAATTGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM83B PubMed Links

Gene Details

Gene
FAM83B
Gene Name
family with sequence similarity 83 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010872.2 Intron NP_001010872.1
XM_006715022.3 Intron XP_006715085.1
XM_011514394.2 Intron XP_011512696.1
XM_011514395.2 Intron XP_011512697.1
XM_017010478.1 Intron XP_016865967.1

View Full Product Details