Product Details

SNP ID
rs61732378
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:63324362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATATACAGAGTCTTTTGACAGAC[A/T]GTCCCACTGCTCTGGAGTCCTTTAT
Phenotype
MIM: 603263
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CA12 PubMed Links

Gene Details

Gene
CA12
Gene Name
carbonic anhydrase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001218.4 3345 UTR 3 NP_001209.1
NM_001293642.1 3345 UTR 3 NP_001280571.1
NM_206925.2 3345 UTR 3 NP_996808.1

View Full Product Details