Product Details

SNP ID
rs34704344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:63323472 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTATCCCCATAACAGGGAGCTACA[C/G]AGTGCTTTGCAAATAGTAATGCTTT
Phenotype
MIM: 603263
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CA12 PubMed Links

Gene Details

Gene
CA12
Gene Name
carbonic anhydrase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001218.4 Intron NP_001209.1
NM_001293642.1 Intron NP_001280571.1
NM_206925.2 Intron NP_996808.1

View Full Product Details