Product Details
- SNP ID
-
rs12102486
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:69430571 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTGGATTGCAGTGTAAAATTATTT[C/T]CTTACTGGGTATTACAAGTGAAAAT
- Phenotype
-
MIM: 611964
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CYB5B
PubMed Links
Gene Details
- Gene
- CYB5B
- Gene Name
- cytochrome b5 type B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_030579.2 |
|
Intron |
|
|
NP_085056.2 |
View Full Product Details