Product Details

SNP ID
rs10927864
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:15839166 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGGGAACCCTTAGCTAAAGTGAC[C/G]CAGATGCTGACTTCCAAAAGTCATT
Phenotype
MIM: 613484
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FLJ37453 PubMed Links

Gene Details

Gene
FLJ37453
Gene Name
uncharacterized LOC729614
There are no transcripts associated with this gene.

Gene
SPEN
Gene Name
spen family transcriptional repressor
There are no transcripts associated with this gene.

View Full Product Details