Product Details
- SNP ID
-
rs11586911
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
21
- Location
-
Chr.1:15834894 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CGAGCCGAGAAGCGAAGAACAAAGC[A/G]GCCGCGTTAGCCCGCCGCCCCGAGG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ37453
PubMed Links
Gene Details
- Gene
- FLJ37453
- Gene Name
- uncharacterized LOC729614
There are no transcripts associated with this gene.
View Full Product Details