Product Details
- SNP ID
-
rs4661342
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
25
- Location
-
Chr.1:15834278 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- CAAAAGAAGTTAAATGGGAGATATT[C/G]TCAACAGGCAAAAATAAGATATTCC
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ37453
PubMed Links
Gene Details
- Gene
- FLJ37453
- Gene Name
- uncharacterized LOC729614
There are no transcripts associated with this gene.
View Full Product Details