Product Details

SNP ID
rs2718689
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99518922 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGGCTTCACATCCTCAATCTAAA[C/T]TAGATTTTCTTATGGTATTTTTTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C4orf17 PubMed Links

Gene Details

Gene
C4orf17
Gene Name
chromosome 4 open reading frame 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032149.2 Intron NP_115525.2
XM_011532315.1 Intron XP_011530617.1

View Full Product Details