Product Details
- SNP ID
-
rs4452753
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:55808485 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTTAAGAATTCCCTTTTTTCTTAG[A/G]AATATATATACACTGCTGGTGGGAA
- Phenotype
-
MIM: 612140
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SEPT14
PubMed Links
Gene Details
- Gene
- SEPT14
- Gene Name
- septin 14
View Full Product Details