Product Details

SNP ID
rs7553368
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:161770520 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGCCTTCTTCTTGTGTCCTCACA[C/T]GGCACTGAGAGAGAGAATCTTCTTC
Phenotype
MIM: 605537
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATF6 PubMed Links

Gene Details

Gene
ATF6
Gene Name
activating transcription factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007348.3 Intron NP_031374.2
XM_006711224.1 Intron XP_006711287.1
XM_011509308.1 Intron XP_011507610.1
XM_011509309.1 Intron XP_011507611.1
XM_011509310.2 Intron XP_011507612.1

View Full Product Details