Product Details

SNP ID
rs164122
Assay Type
Validated
NCBI dbSNP Submissions
79
Location
Chr.1:162398567 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATTGAGCAAAGTGTAACAGTATACG[C/T]TTTGGGGAGGGCTTGAAGGGCTAGA
Phenotype
MIM: 608510
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SH2D1B PubMed Links

Gene Details

Gene
SH2D1B
Gene Name
SH2 domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053282.4 Intron NP_444512.2

View Full Product Details