Product Details

SNP ID
rs13152799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:146639259 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCTCGCCGGGCTCCTCGGCTCCCA[C/T]CGCGCCCTCGGCCAGCTCCCCCAGC
Phenotype
MIM: 113725
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POU4F2 PubMed Links

Gene Details

Gene
POU4F2
Gene Name
POU class 4 homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004575.2 367 Missense Mutation ACC,ATC T40I NP_004566.2

View Full Product Details