Product Details

SNP ID
rs35432967
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:58114680 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GTGCTGGAAGGAGACCTAAGATCTT[A/G]CGATATAAAGGCACCCCAAAGTGTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFAP20 PubMed Links

Gene Details

Gene
CFAP20
Gene Name
cilia and flagella associated protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013242.2 Intron NP_037374.1

View Full Product Details