Product Details

SNP ID
rs3821919
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:111732756 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTCTTGTAATAACAAAATGAGCA[C/G]CATATACAGCCTCGTCACCAGAGTG
Phenotype
MIM: 610298 MIM: 617015
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PHLDB2 PubMed Links

Gene Details

Gene
PHLDB2
Gene Name
pleckstrin homology like domain family B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134437.1 Intron NP_001127909.1
NM_001134438.1 Intron NP_001127910.1
NM_001134439.1 Intron NP_001127911.1
NM_145753.2 Intron NP_665696.1
Gene
PLCXD2
Gene Name
phosphatidylinositol specific phospholipase C X domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185106.1 Intron NP_001172035.1
NM_153268.3 Intron NP_695000.1

View Full Product Details