Product Details
- SNP ID
-
rs7130954
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:123362601 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGTGATGTACTACCTACCTACTC[C/T]CCCACCCCATGAGTTGCTGAGCTCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GRAMD1B
PubMed Links
Gene Details
- Gene
- GRAMD1B
- Gene Name
- GRAM domain containing 1B
View Full Product Details