Product Details

SNP ID
rs35545442
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67824158 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGCCTTGAGGTCACCAAGGCCC[C/T]TCAGCCATTCTGCACAGACTGTTCC
Phenotype
MIM: 611510 MIM: 607720
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CENPT PubMed Links

Gene Details

Gene
CENPT
Gene Name
centromere protein T
There are no transcripts associated with this gene.

Gene
TSNAXIP1
Gene Name
translin associated factor X interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288990.1 Intron NP_001275919.1
NM_001288991.1 Intron NP_001275920.1
NM_001288992.1 Intron NP_001275921.1
NM_001288993.1 Intron NP_001275922.1
NM_001288994.1 Intron NP_001275923.1
NM_018430.3 Intron NP_060900.2
XM_005256049.4 Intron XP_005256106.1
XM_011523230.2 Intron XP_011521532.1
XM_011523231.2 Intron XP_011521533.1
XM_011523232.2 Intron XP_011521534.1
XM_011523233.2 Intron XP_011521535.1
XM_011523234.2 Intron XP_011521536.1
XM_011523235.1 Intron XP_011521537.1
XM_011523236.2 Intron XP_011521538.1
XM_011523237.2 Intron XP_011521539.1
XM_011523238.1 Intron XP_011521540.1
XM_011523239.1 Intron XP_011521541.1
XM_011523240.1 Intron XP_011521542.1
XM_011523241.1 Intron XP_011521543.1
XM_011523242.1 Intron XP_011521544.1
XM_011523243.1 Intron XP_011521545.1
XM_017023445.1 Intron XP_016878934.1
XM_017023446.1 Intron XP_016878935.1
XM_017023447.1 Intron XP_016878936.1
XM_017023448.1 Intron XP_016878937.1
XM_017023449.1 Intron XP_016878938.1
XM_017023450.1 Intron XP_016878939.1
XM_017023451.1 Intron XP_016878940.1
XM_017023452.1 Intron XP_016878941.1
XM_017023453.1 Intron XP_016878942.1
XM_017023454.1 Intron XP_016878943.1
XM_017023455.1 Intron XP_016878944.1

View Full Product Details