Product Details
- SNP ID
-
rs10403256
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:51312636 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GTGCTGGGCTGGGGTCTTGGATTCC[C/G]AAGCTCTGATTGAATGTCTAGGTTC
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
IGLON5
PubMed Links
Gene Details
- Gene
- IGLON5
- Gene Name
- IgLON family member 5
View Full Product Details