Product Details

SNP ID
rs11710365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:119598904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACTGCATGCTCTCCTTGCACTCA[C/T]GGGTGGTTTTTCAGAGGTGCTGTTG
Phenotype
MIM: 603081 MIM: 607460
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADPRH PubMed Links

Gene Details

Gene
ADPRH
Gene Name
ADP-ribosylarginine hydrolase
There are no transcripts associated with this gene.

Gene
PLA1A
Gene Name
phospholipase A1 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206960.1 Intron NP_001193889.1
NM_001206961.1 Intron NP_001193890.1
NM_001293225.1 Intron NP_001280154.1
NM_015900.3 Intron NP_056984.1
XM_017006572.1 Intron XP_016862061.1

View Full Product Details