Product Details

SNP ID
rs8053890
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:84055363 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGAACCCAGAGAAACTCCTCTTGG[C/T]AGCTATAAACTCTTTGGAAGAGTAC
Phenotype
MIM: 603355
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MBTPS1 PubMed Links

Gene Details

Gene
MBTPS1
Gene Name
membrane bound transcription factor peptidase, site 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003791.3 Intron NP_003782.1
XM_017023816.1 Intron XP_016879305.1
XM_017023817.1 Intron XP_016879306.1

View Full Product Details