Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000985.4 | Intron | NP_000976.1 | |||
NM_001035006.2 | Intron | NP_001030178.1 | |||
NM_001199340.1 | Intron | NP_001186269.1 | |||
NM_001199341.1 | Intron | NP_001186270.1 | |||
NM_001199342.1 | Intron | NP_001186271.1 | |||
NM_001199343.1 | Intron | NP_001186272.1 | |||
NM_001199344.1 | Intron | NP_001186273.1 | |||
NM_001199345.1 | Intron | NP_001186274.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001199355.1 | Intron | NP_001186284.1 | |||
NM_001199356.1 | Intron | NP_001186285.1 |