Product Details
- SNP ID
-
rs7842666
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:27772635 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GACTCGCGGCGGCCGCCTGGCCCCC[A/G]GAACTCCTCCGTGCACTTCCGGCGG
- Phenotype
-
MIM: 609353
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC25
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114445285] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC25
- Gene Name
- coiled-coil domain containing 25
- Gene
- ESCO2
- Gene Name
- establishment of sister chromatid cohesion N-acetyltransferase 2
View Full Product Details