Product Details

SNP ID
rs3733230
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:82630192 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGGCATGATTTATAAGGTGGGCTGG[C/G]CATAGGGCTCAGGACACAAAACGAG
Phenotype
MIM: 608370
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00575 PubMed Links

Gene Details

Gene
LINC00575
Gene Name
long intergenic non-protein coding RNA 575
There are no transcripts associated with this gene.

Gene
SCD5
Gene Name
stearoyl-CoA desaturase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037582.2 2448 UTR 3 NP_001032671.2
NM_024906.2 2448 Intron NP_079182.2

View Full Product Details