Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_144679.2 | 3280 | UTR 3 | NP_653280.1 | ||
XM_005257066.2 | 3280 | UTR 3 | XP_005257123.1 | ||
XM_005257067.2 | 3280 | UTR 3 | XP_005257124.1 | ||
XM_006721709.3 | 3280 | UTR 3 | XP_006721772.1 | ||
XM_006721712.2 | 3280 | UTR 3 | XP_006721775.1 | ||
XM_011524355.1 | 3280 | UTR 3 | XP_011522657.1 | ||
XM_011524356.1 | 3280 | UTR 3 | XP_011522658.1 | ||
XM_011524357.1 | 3280 | UTR 3 | XP_011522659.1 | ||
XM_011524358.2 | 3280 | UTR 3 | XP_011522660.1 | ||
XM_017024202.1 | 3280 | UTR 3 | XP_016879691.1 | ||
XM_017024203.1 | 3280 | UTR 3 | XP_016879692.1 | ||
XM_017024204.1 | 3280 | UTR 3 | XP_016879693.1 |