Product Details

SNP ID
rs948971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:77225788 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTACGTGCCCATCCTGGAGGTGG[C/T]ATGGGGCAACACATGGCACTCCGTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GDPD4 PubMed Links

Gene Details

Gene
GDPD4
Gene Name
glycerophosphodiester phosphodiesterase domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182833.1 5151 Intron NP_878253.1
XM_011544826.1 5151 Intron XP_011543128.1
XM_011544833.1 5151 Intron XP_011543135.1
XM_011544834.1 5151 Intron XP_011543136.1
XM_017017348.1 5151 UTR 3 XP_016872837.1
XM_017017349.1 5151 UTR 3 XP_016872838.1
XM_017017350.1 5151 UTR 3 XP_016872839.1
XM_017017351.1 5151 UTR 3 XP_016872840.1

View Full Product Details