Product Details

SNP ID
rs11593747
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:59792628 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTCTGGGCCAAGCAAAAATTGCA[C/T]ACTGCTGCTGAAATACCAAATACCA
Phenotype
MIM: 601985
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC6 PubMed Links

Gene Details

Gene
CCDC6
Gene Name
coiled-coil domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005436.4 1946 UTR 3 NP_005427.2

View Full Product Details