Product Details

SNP ID
rs6801075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:126393456 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTCTCATTAGAGCACCTTGTATT[C/T]AGATAAACCATTACAAAAACACTTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC37-AS1 PubMed Links

Gene Details

Gene
CCDC37-AS1
Gene Name
CCDC37 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
CFAP100
Gene Name
cilia and flagella associated protein 100
There are no transcripts associated with this gene.

View Full Product Details