Product Details

SNP ID
rs6438995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:126294529 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAACGTCAATAATTAAGCCATACA[A/C]CAGTTGATAGCCTGTGGGGCAGGAG
Phenotype
MIM: 606465
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KLF15 PubMed Links

Gene Details

Gene
KLF15
Gene Name
Kruppel like factor 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014079.3 Intron NP_054798.1
XM_005247400.3 Intron XP_005247457.1
XM_011512743.2 Intron XP_011511045.1

View Full Product Details