Product Details

SNP ID
rs17354606
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:58251562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTTCATTTCCCTGATGTAATGTT[C/T]ATCTGGATCCACTGACATGTCCCCT
Phenotype
MIM: 616966
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABHD6 PubMed Links

Gene Details

Gene
ABHD6
Gene Name
abhydrolase domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320126.1 Intron NP_001307055.1
NM_020676.6 Intron NP_065727.4
XM_005265334.3 Intron XP_005265391.1
XM_005265335.3 Intron XP_005265392.1
XM_017006930.1 Intron XP_016862419.1

View Full Product Details