Product Details

SNP ID
rs10143899
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:31562923 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCGCCCTGCCTACTAGCTCATTTTT[C/T]CCCATTTTTATCAGAAGGGAATTCT
Phenotype
MIM: 613621
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUBPL PubMed Links

Gene Details

Gene
NUBPL
Gene Name
nucleotide binding protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201573.1 Intron NP_001188502.1
NM_001201574.1 Intron NP_001188503.1
NM_025152.2 Intron NP_079428.2
XM_011537181.2 Intron XP_011535483.1
XM_011537182.2 Intron XP_011535484.1
XM_011537183.2 Intron XP_011535485.1
XM_011537184.2 Intron XP_011535486.1
XM_017021664.1 Intron XP_016877153.1
XM_017021665.1 Intron XP_016877154.1
XM_017021666.1 Intron XP_016877155.1
XM_017021667.1 Intron XP_016877156.1

View Full Product Details