Product Details

SNP ID
rs10499059
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:110986235 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AATTCTAATCTGAAAGCTCCGTGTA[A/G]TCATTGAAAGATATTAAGCAAGAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPF2 PubMed Links

Gene Details

Gene
RPF2
Gene Name
ribosome production factor 2 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289111.1 Intron NP_001276040.1
NM_032194.2 Intron NP_115570.1
XM_017011354.1 Intron XP_016866843.1

View Full Product Details