Product Details

SNP ID
rs7524537
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:162643332 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGAGTTGCCAACATCTACAAAATA[C/T]ATTCTAACAAAGATGCTCAGAATGT
Phenotype
MIM: 191311
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDR2 PubMed Links

Gene Details

Gene
DDR2
Gene Name
discoidin domain receptor tyrosine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014796.1 Intron NP_001014796.1
NM_006182.2 Intron NP_006173.2
XM_006711344.3 Intron XP_006711407.1
XM_011509586.2 Intron XP_011507888.1
XM_011509587.2 Intron XP_011507889.1
XM_011509588.2 Intron XP_011507890.1

View Full Product Details