Product Details
- SNP ID
-
rs6793304
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:20165531 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACAGATTCAAGAAGAATTAGAGAAC[A/G]TGTAAGACTTATAATTATTAAAAAA
- Phenotype
-
MIM: 609168
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SGO1
PubMed Links
Gene Details
- Gene
- SGO1
- Gene Name
- shugoshin 1
- Gene
- SGO1-AS1
- Gene Name
- SGO1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details