Product Details
- SNP ID
-
rs34673412
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:20169618 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCTTTCATTGGTTGGTCAAAAATA[C/T]GCAAAAACCCTAAATATTCACACAT
- Phenotype
-
MIM: 609168
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SGO1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112048159] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SGO1
- Gene Name
- shugoshin 1
- Gene
- SGO1-AS1
- Gene Name
- SGO1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details