Product Details

SNP ID
rs12001617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33293098 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATTTTTCACTGTATGTTTCTATG[A/G]TACCCACTGCCCAAGCTCTGTTAGT
Phenotype
MIM: 603255
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NFX1 PubMed Links

Gene Details

Gene
NFX1
Gene Name
nuclear transcription factor, X-box binding 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318758.1 Intron NP_001305687.1
NM_002504.5 Intron NP_002495.2
NM_147134.3 Intron NP_667345.1

View Full Product Details