Product Details

SNP ID
rs2304445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5033603 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAAGCTCTGCACAGAAGGCAGCA[C/T]GCCATTGGTCACGGCACTGGTGAAG
Phenotype
MIM: 603060 MIM: 607883
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF1C PubMed Links

Gene Details

Gene
KIF1C
Gene Name
kinesin family member 1C
There are no transcripts associated with this gene.

Gene
SLC52A1
Gene Name
solute carrier family 52 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104577.1 1599 Missense Mutation ATG,GTG M296V NP_001098047.1
NM_017986.3 1599 Missense Mutation ATG,GTG M296V NP_060456.3
XM_011523951.1 1599 Missense Mutation ATG,GTG M296V XP_011522253.1

View Full Product Details