Product Details
- SNP ID
-
rs12459671
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:3884724 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCCCACGTTCAAACTCTGAACCCCA[C/G]TGTGCATCCTAAGCTCTCTGTGCCT
- Phenotype
-
MIM: 608179
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ATCAY
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78332543] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATCAY
- Gene Name
- ATCAY, caytaxin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033064.4 |
|
Intron |
|
|
NP_149053.1 |
View Full Product Details