Product Details

SNP ID
rs2292274
Assay Type
Validated
NCBI dbSNP Submissions
53
Location
Chr.1:163322250 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
CCCGTCGTTACTTTTTATTTGATGT[C/T]AGTCCCAGAAACAACCAGAGCTCAG
Phenotype
MIM: 611772 MIM: 603276
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUF2 PubMed Links

Gene Details

Gene
NUF2
Gene Name
NUF2, NDC80 kinetochore complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031423.3 Intron NP_113611.2
NM_145697.2 Intron NP_663735.2
XM_011510036.1 Intron XP_011508338.1
Gene
RGS5
Gene Name
regulator of G-protein signaling 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195303.2 Intron NP_001182232.1
NM_001254748.1 Intron NP_001241677.1
NM_001254749.1 Intron NP_001241678.1
NM_003617.3 Intron NP_003608.1

View Full Product Details