Product Details
- SNP ID
-
hCV1568115
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:36089672 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACCTAGAGTGAGCTGGAGAGTGAA[C/G]AACATACCCCACTGGGAAGTAAGCA
- Phenotype
-
MIM: 182283
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CCL3
PubMed Links
Gene Details
- Gene
- CCL3
- Gene Name
- C-C motif chemokine ligand 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002983.2 |
|
Intron |
|
|
NP_002974.1 |
View Full Product Details