Product Details
- SNP ID
-
rs949646
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:39603866 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATTTAGTCCGGAGCTTCTCACTCT[G/T]TAACATGCACACAAATCACCTGCGG
- Phenotype
-
MIM: 615795
MIM: 188060
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FSIP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78120737,rs79544755] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FSIP1
- Gene Name
- fibrous sheath interacting protein 1
- Gene
- THBS1
- Gene Name
- thrombospondin 1
There are no transcripts associated with this gene.
View Full Product Details